Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSIG4 Antibody (005), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290065JF646
Description
VSIG4 Monoclonal specifically detects VSIG4 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
VSIG4 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
11326 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
5 | |
ELISA, Sandwich ELISA Detection | |
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human VSIG4 (Q9Y279-1; Met 1-Pro 283). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction