Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSIG4 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299947MFV610
Description
VSIG4 Polyclonal antibody specifically detects VSIG4 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
VSIG4 | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily | |
Produced in rabbits immunized with purified, recombinant Human VSIG4 (Uniprot#: Q9Y279-1; Met1-Pro283) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
11326 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction