Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSIG4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$272.00
Specifications
Antigen | VSIG4 |
---|---|
Dilution | ELISA 1:5000-1:10000, Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 |
Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
VSIG4 Polyclonal antibody specifically detects VSIG4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
VSIG4 | |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily | |
Produced in rabbits immunized with purified, recombinant Human VSIG4 (Q9Y279-1; Met1-Pro283). | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
ELISA 1:5000-1:10000, Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
11326 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title