Learn More
VSIG4 Monoclonal Antibody (JAV4), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$180.00 - $438.00
Specifications
Antigen | VSIG4 |
---|---|
Clone | JAV4 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
62-575-741
![]() |
Invitrogen™
62575741 |
25 Tests |
Each for $180.00
|
|
|||||
62-575-742
![]() |
Invitrogen™
62575742 |
100 Tests |
Each for $438.00
|
![]() |
|||||
Description
This JAV4 monoclonal antibody recognizes human V-Set and Immunoglobulin domain containing 4 (VSIG4), also known as Complement Receptor of the Immunoglobulin superfamily (CRIg) or Z39Ig. There are two variants of human VSIG4 that result from alternative sp
T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.Specifications
VSIG4 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG | |
VSIG4 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
VSIG4 |
JAV4 | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
Human | |
Q9Y279 | |
11326 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.