Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSIG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552018
Description
Immunogen sequence: PPISYIWYKQ QTNNQEPIKV ATLSTLLFKP AVIADSGSYF CTAKGQVGSE QHSDIVKFVV KDSSKLLKTK TEAPTTMTYP LKATSTVKQS WDWTTDMDGY L Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 25%.
T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
Specifications
VSIG4 | |
Polyclonal | |
Unconjugated | |
VSIG4 | |
A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11326 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y279 | |
VSIG4 | |
Recombinant protein corresponding to Human VSIG4. Recombinant protein control fragment (Product #RP-92217). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction