Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSIG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581117 Supplier Invitrogen™ Supplier No. PA581117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human VSIG4.

T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSIG4
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene VSIG4
Gene Accession No. Q9Y279
Gene Alias A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG
Gene Symbols VSIG4
Host Species Rabbit
Immunogen Recombinant Human VSIG4 Protein (Met1-Pro283).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11326
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.