Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSIG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542027
Description
Peptide sequence: VPGDVSLQLS TLEMDDRSHY TCEVTWQTPD GNQVVRDKIT ELRVQKLSVS Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 92%; Horse: 91%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 82%; Rat: 86%.
T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
Specifications
VSIG4 | |
Polyclonal | |
Unconjugated | |
VSIG4 | |
A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG | |
Rabbit | |
Protein A | |
RUO | |
11326 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y279 | |
VSIG4 | |
Synthetic peptide directed towards the N-terminal of human VSIG4 (aa 92-141). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction