Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSIG8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554724
Description
Immunogen sequence: LDPEDYGPNG LDIEWMQVNS DPAHHRENVF LSYQDKRINH GSLPHLQQRV RFAASDPSQY DASINLMNLQ VSDTATYECR VKKTTMATRK VIVTVQARPA VPMCWTEGHM TYGNDVVLKC YASGGSQPLS YKWAK Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
VSIG8 contains 2 Ig-like V-type (immunoglobulin-like) domains. VSIG8 is single-pass type I membrane protein. The function of the VSIG8 protein remains unknown.
Specifications
VSIG8 | |
Polyclonal | |
Unconjugated | |
VSIG8 | |
A030011M19; C1orf204; EG240916; LOC100727929; LOW QUALITY PROTEIN: V-set and immunoglobulin domain-containing protein 8; RGD1562464; V-set and immunoglobulin domain containing 8; V-set and immunoglobulin domain containing 8; V-set and immunoglobulin domain-containing protein 8; V-set and immunoglobulin domain-containing protein 8; Vsig8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
391123 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P0DPA2 | |
VSIG8 | |
Recombinant protein corresponding to Human VSIG8. Recombinant protein control fragment (Product #RP-90576). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction