Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSIG8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569338 Supplier Invitrogen™ Supplier No. PA569338
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

VSIG8 contains 2 Ig-like V-type (immunoglobulin-like) domains. VSIG8 is single-pass type I membrane protein. The function of the VSIG8 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSIG8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene VSIG8
Gene Accession No. P0DPA2
Gene Alias A030011M19; C1orf204; EG240916; LOC100727929; LOW QUALITY PROTEIN: V-set and immunoglobulin domain-containing protein 8; RGD1562464; V-set and immunoglobulin domain containing 8; V-set and immunoglobulin domain containing 8; V-set and immunoglobulin domain-containing protein 8; V-set and immunoglobulin domain-containing protein 8; Vsig8
Gene Symbols VSIG8
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human VSIG8.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 391123
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.