Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSIG8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551157
Description
The antibody detects endogenous levels of total VSIG8 protein.
VSIG8 contains 2 Ig-like V-type (immunoglobulin-like) domains. VSIG8 is single-pass type I membrane protein. The function of the VSIG8 protein remains unknown.
Specifications
VSIG8 | |
Polyclonal | |
Unconjugated | |
VSIG8 | |
A030011M19; C1orf204; EG240916; LOC100727929; LOW QUALITY PROTEIN: V-set and immunoglobulin domain-containing protein 8; RGD1562464; V-set and immunoglobulin domain containing 8; V-set and immunoglobulin domain containing 8; V-set and immunoglobulin domain-containing protein 8; V-set and immunoglobulin domain-containing protein 8; Vsig8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
240916, 391123 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P0DPA2, Q6P3A4 | |
VSIG8 | |
Fusion protein of human VSIG8. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction