Learn More
Invitrogen™ VSNL1 Monoclonal Antibody (2D11)
Description
VSNL1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Specifications
Specifications
| Antigen | VSNL1 |
| Applications | Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2D11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 5mM sodium azide |
| Gene | VSNL1 |
| Gene Accession No. | P62760, P62761, P62762 |
| Gene Alias | 21 kDa CABP; hippocalcin-like protein 3; HLP3; HPCAL3; HUVISL1; Neural visinin-like protein 1; neurocalcin alpha; Neurocalcin-alpha; NVL-1; Nvp1; NVP-1; Ratnvp1; VILIP; VILIP-1; visinin like 1; visinin-like 1; visinin-like protein 1; Visl1; VLP-1; Vnsl1; VSNL1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.