Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSNL1 Monoclonal Antibody (3A9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547310
Description
VSNL1 Monoclonal Antibody for Western Blot, ICC/IF, IHC
This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
Specifications
VSNL1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 5mM sodium azide | |
P62760, P62761, P62762, P62763 | |
VSNL1 | |
Recombinant full length human protein. | |
500 μL | |
Primary | |
Human, Mouse, Rat, Bovine, Pig | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
3A9 | |
Unconjugated | |
VSNL1 | |
21 kDa CABP; hippocalcin-like protein 3; HLP3; HPCAL3; HUVISL1; Neural visinin-like protein 1; neurocalcin alpha; Neurocalcin-alpha; NVL-1; Nvp1; NVP-1; Ratnvp1; VILIP; VILIP-1; visinin like 1; visinin-like 1; visinin-like protein 1; Visl1; VLP-1; Vnsl1; VSNL1 | |
Mouse | |
Protein A/G | |
RUO | |
100517707, 24877, 26950, 282124, 7447 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction