Learn More
VSNL1 Mouse anti-Human, Clone: OTI3B9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802075

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.Specifications
VSNL1 | |
Monoclonal | |
Unconjugated | |
VSNL1 | |
HLP3, HPCAL3, HUVISL1, VILIP, VILIP-1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7447 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3B9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P62760 | |
VSNL1 | |
Human recombit protein fragment corresponding to amino acids 2-191 of human VSNL1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.