Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSNL1 Recombinant Rabbit Monoclonal Antibody (0R1V3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551351
Description
Immunogen sequence: GKQNSKLAPE VMEDLVKSTE FNEHELKQWY KGFLKDCPSG RLNLEEFQQL YVKFFPYGDA SKFAQHAFRT FDKNGDGTID FREFICALSI TSRGSFEQKL NWAFNMYDLD GDGKITRVEM LEIIEAIYKM VGTVIMMKMN EDGLTPEQRV DKIFSKMDKN KDDQITLDEF KEAAKSDPSI VLLLQCDIQK.
This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
Specifications
VSNL1 | |
Recombinant Monoclonal | |
0.53 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P62760, P62761, P62762 | |
VSNL1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 2-191 of human VSNL1/VILIP-1 (NP_003376.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Western Blot | |
0R1V3 | |
Unconjugated | |
VSNL1 | |
21 kDa CABP; hippocalcin-like protein 3; HLP3; HPCAL3; HUVISL1; Neural visinin-like protein 1; neurocalcin alpha; Neurocalcin-alpha; NVL-1; Nvp1; NVP-1; Ratnvp1; VILIP; VILIP-1; visinin like 1; visinin-like 1; visinin-like protein 1; Visl1; VLP-1; Vnsl1; VSNL1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
24877, 26950, 7447 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction