Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSP15a Rabbit anti-Yeast, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | VSP15a |
---|---|
Concentration | 0.13 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5899
![]() ![]() |
Proteintech Group Inc
510901AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody recognizes the SPAPB1A10 protein of Yeast.
Specifications
VSP15a | |
ELISA | |
Unconjugated | |
Rabbit | |
Yeast | |
SPAPB1A10, VSP15a | |
SPAPB1A10.05 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
2543600 | |
VSP15a Fusion Protein Ag0724 | |
Primary | |
-20°C | |
SPAPB1A10.05 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title