Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSTM1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305911PE
Description
VSTM1 Polyclonal antibody specifically detects VSTM1 in Human samples. It is validated for ELISASpecifications
VSTM1 | |
Polyclonal | |
ELISA | |
LAIR homolog, MGC119160, MGC119161, OSCAR-like transcript-1, signal inhibitory receptor on leukocytes-1, SIRL1, UNQ3033, V-set and transmembrane domain containing 1, V-set and transmembrane domain-containing protein 1 | |
Produced in rabbits immunized with purified, recombinant Human VSTM1 (Uniprot#: Q6UX27-16; Met1-Thr135) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
284415 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction