Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSTM1 (SIRL-1) Mouse anti-Human, Alexa Fluor™ 647, Clone: 1A5/SIRL-1 (also known as 1A5), BD Pharmingen™
Mouse Monoclonal Antibody
$128.00 - $313.50
Specifications
Antigen | VSTM1 (SIRL-1) |
---|---|
Clone | 1A5/SIRL-1 (also known as 1A5) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Description
VSTM1 (SIRL-1) Monoclonal antibody specifically detects VSTM1 (SIRL-1) in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
VSTM1 (SIRL-1) | |
Flow Cytometry | |
Alexa Fluor 647 | |
Mouse | |
Human | |
VSTM1; SIRL1; SIRL-1 | |
Purified SIRL-1-hIg Fusion Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
1A5/SIRL-1 (also known as 1A5) | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
284415 | |
IgG1 κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title