Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSTM2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | VSTM2A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
VSTM2A Polyclonal specifically detects VSTM2A in Human samples. It is validated for Western Blot.Specifications
| VSTM2A | |
| Polyclonal | |
| Rabbit | |
| NP_872352 | |
| 222008 | |
| Synthetic peptide directed towards the middle region of human VSTM2AThe immunogen for this antibody is VSTM2A. Peptide sequence DISHKLQISKVRKKDEGLYECRVTDANYGELQEHKAQAYLKVNANSHARR. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| MGC33530, V-set and transmembrane domain containing 2, V-set and transmembrane domain containing 2A, V-set and transmembrane domain-containing protein 2A, VSTM2 | |
| VSTM2A | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title