Learn More
Invitrogen™ VSTM2A Polyclonal Antibody

Description
VSTM2A Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | VSTM2A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | VSTM2A |
| Gene Accession No. | Q8R0A6, Q8TAG5 |
| Gene Alias | hypothetical protein MGC33530; V-set and transmembrane domain containing 2; V-set and transmembrane domain containing 2A; V-set and transmembrane domain-containing protein 2A; Vstm2; VSTM2A |
| Gene Symbols | VSTM2A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.