Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSTM2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554990 Supplier Invitrogen™ Supplier No. PA554990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KFTEFPRNVT ATEGQNVEMS CAFQSGSASV YLEIQWWFLR GPEDLDPGAE GAGAQVELLP DRDPDSDGTK ISTVK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 71%.

Plays a role in the regulation of the early stage of white and brown preadipocyte cell differentiation. Promotes adipogenic commitment of preadipocytes by increasing gene expression of the transcription factor PPARG in a BMP4-dependent signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSTM2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VSTM2A
Gene Accession No. Q8TAG5
Gene Alias hypothetical protein MGC33530; V-set and transmembrane domain containing 2; V-set and transmembrane domain containing 2A; V-set and transmembrane domain-containing protein 2A; Vstm2; VSTM2A
Gene Symbols VSTM2A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VSTM2A. Recombinant protein control fragment (Product #RP-92865).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222008
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.