Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSTM2L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560529
Description
Immunogen sequence: QWWYVRSHRD WTDKQAWASN QLKASQQEDA GKEATKISVV KVVGSNISHK LRLSRVKPTD EGTYEC Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Specifications
VSTM2L | |
Polyclonal | |
Unconjugated | |
VSTM2L | |
C20orf102; dJ1118M15.2; V-set and transmembrane domain containing 2 like; V-set and transmembrane domain-containing protein 2-like protein; VSTM2L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
128434 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96N03 | |
VSTM2L | |
Recombinant protein corresponding to Human VSTM2L. Recombinant protein control fragment (Product #RP-99437). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction