Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSTM4 Antibody (2694A), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10767H
Description
VSTM4 Monoclonal antibody specifically detects VSTM4 in Human samples. It is validated for Flow CytometrySpecifications
VSTM4 | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
0.1 mL | |
Primary | |
Human | |
IgG |
Flow Cytometry | |
2694A | |
Flow Cytometry | |
FLJ31737, hypothetical protein LOC196740, V-set and transmembrane domain containing 4 | |
Chinese Hamster Ovary cell line CHO-derived human VSTM4, Leu24-Tyr180, Accession # Q8IW00-1 | |
RUO | |
196740 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction