Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSTM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553572
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553572 Supplier Invitrogen™ Supplier No. PA553572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVRHYLVKCP QNSSGETVTS VTSLAPLQPK KGKRQKEKPD IPPAVPAKAP IAPTFHKPKL LKPQRKVTLP KIAEENLTYA ELELIKPHRA AKGAPTSTVY AQILFE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

Peptide Lv enhances L-type voltage-gated calcium channel (L-VGCC) currents in retinal photoreceptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSTM4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VSTM4
Gene Accession No. Q8IW00
Gene Alias C10orf72; Peptide Lv; V-set and transmembrane domain containing 4; V-set and transmembrane domain-containing protein 4; VSTM4
Gene Symbols VSTM4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VSTM4. Recombinant protein control fragment (Product #RP-90403).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196740
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.