Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSTM5 Antibody (2696A), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10872Y
This item is not returnable.
View return policy
Description
VSTM5 Monoclonal antibody specifically detects VSTM5 in Human samples. It is validated for Flow Cytometry, ImmunohistochemistrySpecifications
| VSTM5 | |
| Monoclonal | |
| DyLight 680 | |
| C11orf90, V-set and transmembrane domain containing 5, V-set and transmembrane domain-containing protein 5 | |
| Human embryonic kidney cell HEK293-derived human VSTM5, Met1-His147, Accession # NP_001138343 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry | |
| 2696A | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 387804 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction