Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSTM5 Antibody (2696A), PE/Cy5.5, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10872PECY55
Description
VSTM5 Monoclonal antibody specifically detects VSTM5 in Human samples. It is validated for Flow CytometrySpecifications
VSTM5 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
387804 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Flow Cytometry | |
2696A | |
Flow Cytometry | |
C11orf90, V-set and transmembrane domain containing 5, V-set and transmembrane domain-containing protein 5 | |
Human embryonic kidney cell HEK293-derived human VSTM5, Met1-His147, Accession # NP_001138343 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction