Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSTM5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584603
Description
Immunogen sequence: NKCAYKFQRK RRHKLKESTT EEIELEDVEC.
VSTM5 is a protein coding gene.
Specifications
| VSTM5 | |
| Polyclonal | |
| Unconjugated | |
| VSTM5 | |
| 2200002K05Rik; C11orf90; C29H11orf90; hypothetical gene supported by BC088439; V-set and transmembrane domain containing 5; V-set and transmembrane domain-containing protein 5; Vstm5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 387804 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A8MXK1 | |
| VSTM5 | |
| Recombinant protein corresponding to Human VSTM5. Recombinant protein control fragment (Product #RP-108490). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction