Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV-G Epitope Tag Rabbit anti-Tag, DyLight™ 680, Polyclonal, Rockland™
SDP

Catalog No. 501992515
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2515 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2515 Supplier Rockland Immunochemicals Supplier No. 600444386
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against the VSV-G epitope tag and is useful in determining its presence in over expressed proteins in various assays. The antibody recognizes the VSV-G epitope tag (Tyr-Thr-Asp-Ile-Glu-Met-Asn-Arg-Leu-Gly-Lys) fused to either the amino- or carboxy- termini of targeted proteins in transfected or transformed cells.

Vesicular stomatitis virus (VSV), an enveloped RNA virus from the Rhabdoviridae family, is released from the plasma membrane of host cells by a process called budding. The glycoprotein (VSV G) contains a domain in its extracellular membrane-proximal stem that appears to be needed for efficient VSV budding: Sequence: YTDIEMNRLGK.

Specifications

Antigen VSV-G Epitope Tag
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate DyLight 680
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2
Gene Alias VSIVgp3; VSV epitope tag; VSV tag; VSVG epitope tag; VSV-G epitope tag; VSVG tag; YTDIEMNRLGK
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.