Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSV-G Polyclonal Antibody, QED Bioscience™

Rabbit Polyclonal Antibody
$482.32 - $3247.60
Specifications
| Antigen | VSV-G |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-7864
![]()
|
Leinco Technologies Inc
1887601100UG |
100 μg |
Each for $482.32
|
|
|||||
50-227-7865
![]()
|
Leinco Technologies Inc
188761MG |
1 mg |
Each for $3,247.60
|
|
|||||
Description
VSV-G Polyclonal antibody specifically detects VSV-G in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
| VSV-G | |
| ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Synthetic peptide YTDIEMNRLGK conjugated to KLH. | |
| Protein A | |
| Antibody |
| 1 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| vesicular stomatitis virus; VSIVgp3; VSV epitope tag; VSV tag; vsvg; VSVG epitope tag; VSV-G epitope tag; VSVG tag; YTDIEMNRLGK | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title