Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV-G Tag Monoclonal Antibody (1000000), Bioss
SDP

Catalog No. 502279711
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9711 Supplier Bioss Supplier No. BSM33006M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VSV-G Tag Monoclonal antibody specifically detects VSV-G Tag in Tag samples. It is validated for ELISA, Western Blot

Specifications

Antigen VSV-G Tag
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1000000
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene Alias vesicular stomatitis virus; VSIVgp3; VSV epitope tag; VSV tag; vsvg; VSVG epitope tag; VSV-G epitope tag; VSVG tag; YTDIEMNRLGK
Host Species Mouse
Immunogen KLH conjugated VSV-G tag
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.