Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV-Glycoprotein Mouse anti-Virus, Clone: 1E9F9, MilliporeSigma™

Catalog No. MABF2321100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2321100 100 μL
MABF232125 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2321100 Supplier MilliporeSigma Supplier No. MABF2321100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VSV-Glycoprotein Monoclonal antibody specifically detects VSV-Glycoprotein in Virus samples. It is validated for ELISA, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralizing, Radioimmune Assays, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSV-Glycoprotein
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 1E9F9
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in PBS without preservatives.
Host Species Mouse
Immunogen Vesicular stomatitis virus, Indiana strain.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.