Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV Matrix Protein M Mouse anti-Virus, Clone: 23H12, MilliporeSigma™

Catalog No. MABF2347100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2347100 100 μg
MABF234725 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2347100 Supplier MilliporeSigma Supplier No. MABF2347100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VSV Matrix Protein M Monoclonal antibody specifically detects VSV Matrix Protein M in Virus samples. It is validated for Immunocytochemistry, Immunofluorescence, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSV Matrix Protein M
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 23H12
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2ak in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Vesicular stomatitis virus, Indiana serotype.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.