Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145225
Description
Immunogen sequence: RQKRSDSVST SDEDSQSEDR NDLKASPTLG KRTKRR Antigen sequence identity: Mouse 67%; Rat 64%.
VSX1 contains a paired-like homeodomain and binds to the core of the locus control region of the red/green visual pigment gene cluster. VSX1 may regulate expression of the cone opsin genes early in development. Mutations in VSX1 can cause posterior polymorphous corneal dystrophy and keratoconus. Alternatively spliced transcript variants encoding different isoforms have been described. The protein encoded by this gene contains a paired-like homeodomain and binds to the core of the locus control region of the red/green visual pigment gene cluster. The encoded protein may regulate expression of the cone opsin genes early in development. Mutations in this gene can cause posterior polymorphous corneal dystrophy and keratoconus. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
VSX1 | |
Polyclonal | |
Unconjugated | |
VSX1 | |
CAASDS; CHX10-like; homeodomain protein RINX; KTCN; KTCN1; PPCD; PPCD1; PPD; Retinal inner nuclear layer homeobox protein; RINX; Transcription factor VSX1; visual system homeobox 1; visual system homeobox 1 homolog (zebrafish); VSX1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
30813 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NZR4 | |
VSX1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 131-166. Recombinant protein control fragment (Product #RP-110239). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction