Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541142 Supplier Invitrogen™ Supplier No. PA541142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FLPPRGPEPA APLAPSRPPP ALGRQKRSDS VSTSDEDSQS EDRNDLKASP Sequence homology: Cow: 93%; Dog: 86%; Horse: 93%; Human: 100%; Mouse: 79%; Pig: 86%; Yeast: 86%.

VSX1 contains a paired-like homeodomain and binds to the core of the locus control region of the red/green visual pigment gene cluster. VSX1 may regulate expression of the cone opsin genes early in development. Mutations in VSX1 can cause posterior polymorphous corneal dystrophy and keratoconus. Alternatively spliced transcript variants encoding different isoforms have been described. The protein encoded by this gene contains a paired-like homeodomain and binds to the core of the locus control region of the red/green visual pigment gene cluster. The encoded protein may regulate expression of the cone opsin genes early in development. Mutations in this gene can cause posterior polymorphous corneal dystrophy and keratoconus. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSX1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene VSX1
Gene Accession No. Q9NZR4
Gene Alias CAASDS; CHX10-like; homeodomain protein RINX; KTCN; KTCN1; PPCD; PPCD1; PPD; Retinal inner nuclear layer homeobox protein; RINX; Transcription factor VSX1; visual system homeobox 1; visual system homeobox 1 homolog (zebrafish); VSX1
Gene Symbols VSX1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human VSX1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30813
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.