Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597243
Description
Immunogen sequence: MTGRDSLSDG RTSSRALVPG GSPRGSRPRG FAITDLLGLE AELPAPAGPG QGSGCEGPAV APCPGPGLDG SSLARGALPL GLGLLCGFGT QPPAAARAPC LLLADVPFLP PRGPEPAAPL APSRPPPALG RQKRSDSVST SDEDSQSEDR NDLKASPTLG KRKKRRHRTV; Positive Samples: A-549; Cellular Location: Nucleus.
VSX1 contains a paired-like homeodomain and binds to the core of the locus control region of the red/green visual pigment gene cluster. VSX1 may regulate expression of the cone opsin genes early in development. Mutations in VSX1 can cause posterior polymorphous corneal dystrophy and keratoconus. Alternatively spliced transcript variants encoding different isoforms have been described. The protein encoded by this gene contains a paired-like homeodomain and binds to the core of the locus control region of the red/green visual pigment gene cluster. The encoded protein may regulate expression of the cone opsin genes early in development. Mutations in this gene can cause posterior polymorphous corneal dystrophy and keratoconus. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
VSX1 | |
Polyclonal | |
Unconjugated | |
VSX1 | |
CAASDS; CHX10-like; homeodomain protein RINX; KTCN; KTCN1; PPCD; PPCD1; PPD; Retinal inner nuclear layer homeobox protein; RINX; Transcription factor VSX1; visual system homeobox 1; visual system homeobox 1 homolog (zebrafish); VSX1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
30813 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NZR4 | |
VSX1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human VSX1 (NP_0554032). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction