Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VTA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP186745
Description
VTA1 Polyclonal specifically detects VTA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VTA1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
C6orf55DRG1, Dopamine-responsive gene 1 protein, DRG-1, FLJ27228, homolog of mouse SKD1-binding protein 1, LIP5HSPC228, LYST-interacting protein 5, My012, SBP1chromosome 6 open reading frame 55, SKD1-binding protein 1, vacuolar protein sorting-associated protein VTA1 homolog, Vps20-associated 1 homolog (S. cerevisiae) | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
VTA1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:FKSIQHHLRTAQEHDKRDPVVAYYCRLYAMQTGMKIDSKTPECRKFLSKLMDQLEALKKQLGDNEA | |
0.1 mL | |
Virology Bacteria and Parasites | |
51534 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction