Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VTA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544649 Supplier Invitrogen™ Supplier No. PA544649
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KKQLGDNEAI TQEIVGCAHL ENYALKMFLY ADNEDRAGRF HKNMIKSFYT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

C6ORF55 encodes a protein involved in trafficking of the multivesicular body, an endosomal compartment involved in sorting membrane proteins for degradation in lysosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VTA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene VTA1
Gene Accession No. Q9NP79
Gene Alias 1110001D18Rik; 1110059P08Rik; AU040813; C6orf55; C85340; chromosome 6 open reading frame 55; dopamine-responsive gene 1 protein; DRG1; DRG-1; homolog of mouse SKD1-binding protein 1; HSPC228; LIP5; lysosomal trafficking regulator interacting protein-5; LYST-interacting protein 5; My012; RGD1305031; SBP1; similar to 1110059P08Rik protein; SKD1 binding protein 1; SKD1-binding protein 1; Vacuolar protein sorting-associated protein VTA1 homolog; vesicle (multivesicular body) trafficking 1; vesicle trafficking 1; Vps20-associated 1 homolog; Vps20-associated 1 homolog (S. cerevisiae); Vta1; wu:fb08c11; zgc:64104
Gene Symbols VTA1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human VTA1 (aa 71-120).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51534
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.