Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VTA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583170
Description
Immunogen sequence: NGETPQAGPV GIEEDNDIEE NEDAGAASLP TQPTQPSSSS TYDPSNMPSG NYTGIQIPPG AHAPANTPAE.
C6ORF55 encodes a protein involved in trafficking of the multivesicular body, an endosomal compartment involved in sorting membrane proteins for degradation in lysosomes.
Specifications
VTA1 | |
Polyclonal | |
Unconjugated | |
VTA1 | |
1110001D18Rik; 1110059P08Rik; AU040813; C6orf55; C85340; chromosome 6 open reading frame 55; dopamine-responsive gene 1 protein; DRG1; DRG-1; homolog of mouse SKD1-binding protein 1; HSPC228; LIP5; lysosomal trafficking regulator interacting protein-5; LYST-interacting protein 5; My012; RGD1305031; SBP1; similar to 1110059P08Rik protein; SKD1 binding protein 1; SKD1-binding protein 1; Vacuolar protein sorting-associated protein VTA1 homolog; vesicle (multivesicular body) trafficking 1; vesicle trafficking 1; Vps20-associated 1 homolog; Vps20-associated 1 homolog (S. cerevisiae); Vta1; wu:fb08c11; zgc:64104 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51534 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NP79 | |
VTA1 | |
Recombinant protein corresponding to Human VTA1. Recombinant protein control fragment (Product #RP-104622). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction