Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VTCN1 Rabbit anti-Human, Unconjugated, Clone: B7H4/2652R, Abnova™

Catalog No. 89339673 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-673 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-673 Supplier Abnova Corporation Supplier No. RAB00658
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

VTCN1 Monoclonal antibody specifically detects VTCN1 in Human samples. It is validated for ELISA, IHC-P, IF, Flow Cyt.

Specifications

Antigen VTCN1
Applications ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone B7H4/2652R
Conjugate Unconjugated
Description V-set domain containing T cell activation inhibitor 1
Gene VTCN1
Gene Alias B7-H4, B7H4, B7S1, B7X, B7h.5, FLJ22418, PRO1291, RP11-229A19.4, VCTN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to human VTCN1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 79679
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.