Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VTI1A Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300340FR
Description
VTI1A Polyclonal specifically detects VTI1A in Human samples. It is validated for ELISA.Specifications
VTI1A | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
143187 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 680 | |
MVti1, vesicle transport through interaction with t-SNAREs homolog 1A, vesicle transport through interaction with t-SNAREs homolog 1A (yeast) | |
Produced in rabbits immunized with purified, recombinant VTI1A (Q96AJ9-2; Met1-Arg187). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction