Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VTI1A Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281876MFV500
Description
VTI1A Polyclonal antibody specifically detects VTI1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| VTI1A | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| MVti1, vesicle transport through interaction with t-SNAREs homolog 1A, vesicle transport through interaction with t-SNAREs homolog 1A (yeast) | |
| VTI1a antibody was raised against a 19 amino acid peptide near the center of human VTI1a. The immunogen is located within amino acids 50 - 100 of VTI1a. Amino Acid Squence: REIPPQSRGMYSNRMRSYK | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 143187 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction