Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VTI1A Antibody (OTI1F4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274862
Description
VTI1A Monoclonal specifically detects VTI1A in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VTI1A | |
Monoclonal | |
LYOPH | |
Western Blot 1:200-4000, Immunohistochemistry 1:150 | |
MVti1, vesicle transport through interaction with t-SNAREs homolog 1A, vesicle transport through interaction with t-SNAREs homolog 1A (yeast) | |
Mouse | |
25 kDa | |
100 μg | |
143187 | |
Human, Mouse, Rat, Canine, Monkey | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI1F4 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
VTI1A | |
Full length human recombinant protein of human VTI1A(NP_660207) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction