Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VTI1A Antibody (OTI1F4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274862B
Description
VTI1A Monoclonal specifically detects VTI1A in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VTI1A | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
VTI1A | |
Full length recombinant protein of human VTI1A (NP_660207) produced in HEK293T cells. | |
0.1 mL | |
143187.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1F4 | |
Western Blot, Immunohistochemistry | |
MVti1, vesicle transport through interaction with t-SNAREs homolog 1A, vesicle transport through interaction with t-SNAREs homolog 1A (yeast) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction