Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VTI1A Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1F4, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA505824

Description
V-SNARE that mediates vesicle transport pathways through interactions with t-SNAREs on the target membrane. These interactions are proposed to mediate aspects of the specificity of vesicle trafficking and to promote fusion of the lipid bilayers. Involved in vesicular transport from the late endosomes to the trans-Golgi network. Along with VAMP7, involved in an non- conventional RAB1-dependent traffic route to the cell surface used by KCNIP1 and KCND2. May be involved in increased cytokine secretion associated with cellular senescence.Specifications
VTI1A | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O89116, Q96AJ9, Q9JI51 | |
Vti1a | |
Full length human recombit protein of human VTI1A produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1F4 | |
Unconjugated | |
VTI1A | |
MMDS3, MVti1, VTI1RP2, Vti1-rp2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
100685447, 143187, 53611, 65277 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction