Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VTI1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562920 Supplier Invitrogen™ Supplier No. PA562920
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSRRLEAGYQ IAVETEQIGQ EMLENLSHDR EKIQRARERL RETDANLGKS SRILTGMLRR GCSVKKQCNL SLAPK Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.

V-SNARE that mediates vesicle transport pathways through interactions with t-SNAREs on the target membrane. These interactions are proposed to mediate aspects of the specificity of vesicle trafficking and to promote fusion of the lipid bilayers. Involved in vesicular transport from the late endosomes to the trans-Golgi network. Along with VAMP7, involved in an non- conventional RAB1-dependent traffic route to the cell surface used by KCNIP1 and KCND2. May be involved in increased cytokine secretion associated with cellular senescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VTI1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VTI1A
Gene Accession No. Q96AJ9
Gene Alias 1110014F16Rik; 1110018K19Rik; 4921537J05Rik; EGK_20072; MMDS3; MVti1; MVti1a; SNARE Vti1a-beta protein; vesicle transport through interaction with t-SNAREs 1 homolog; vesicle transport through interaction with t-SNAREs 1A; vesicle transport through interaction with t-SNAREs homolog 1A; Vesicle transport v-SNARE protein Vti1-like 2; Vti1; VTI1A; Vti1l2; VTI1RP2; Vti1-rp2
Gene Symbols VTI1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VTI1A. Recombinant protein control fragment (Product #RP-101369).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.