Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VTI1A Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549615
Description
V-SNARE that mediates vesicle transport pathways through interactions with t-SNAREs on the target membrane. These interactions are proposed to mediate aspects of the specificity of vesicle trafficking and to promote fusion of the lipid bilayers. Involved in vesicular transport from the late endosomes to the trans-Golgi network. Along with VAMP7, involved in an non- conventional RAB1-dependent traffic route to the cell surface used by KCNIP1 and KCND2. May be involved in increased cytokine secretion associated with cellular senescence.
Specifications
VTI1A | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96AJ9 | |
VTI1A | |
KLH conjugated synthetic peptide between 140-169 amino acids from the C-terminal region of human VTI1A. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
VTI1A | |
Vesicle transport through interaction with t-SNAREs homolog 1A, Vesicle transport v-SNARE protein Vti1-like 2, Vti1-rp2, VTI1A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
143187 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction