Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VTI1B Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP296993F
Description
VTI1B Polyclonal antibody specifically detects VTI1B in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
VTI1B | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
vesicle transport through interaction with t-SNAREs homolog 1B (yeast), Vesicle transport v-SNARE protein Vti1-like 1, vesicle-associated soluble NSF attachment protein receptor, v-SNARE, VTI1, VTI1L, VTI1L1, VTI1-LIKE, Vti1-rp1, VTI2vesicle transport through interaction with t-SNAREs homolog 1B | |
Produced in rabbits immunized with purified, recombinant Human VTI1B (rh VTI1B; NP_006361.1; Ala 2-Leu 208) | |
0.1 mL | |
Signal Transduction | |
10490 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction