Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VTN Mouse anti-Human, Porcine, Rabbit, Clone: 1D2C8, Proteintech
SDP

Catalog No. 501736826 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6826 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6826 Supplier Proteintech Group Inc Supplier No. 663981IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Vitronectin is a plasma glycoprotein that circulates in the blood. Vitronectin is circulating as a mixture of both 75 kDa and 65 kDa forms. Vitronectin is a major cell adhesive glycoprotein and is a common component of extracellular matrix and plasma. It competes effectively with other plasma proteins and is often involved in cell attachment, regulation of blood coagulation and immune responses. It has similar tissue distribution to fibronectin and also its integrin receptor recognises fibronectin.

Specifications

Antigen VTN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1D2C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene VTN
Gene Accession No. P04004, P22458, P48819
Gene Alias S protein, Serum spreading factor, V75, vitronectin, VN, VNT, VTN
Gene Symbols VTN
Host Species Mouse
Immunogen VTN Fusion Protein Ag8443
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009128, 397192, 7448
Target Species Human, Pig, Rabbit
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.