Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 0071063
GREENER_CHOICE

Catalog No. 705710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-571-0 10 mg
70-575-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 70-571-0 Supplier Tocris Bioscience™ Supplier No. 7057/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective activator of SUR1 Kir6.2 channels

VU 0071063 is a selective activator of SUR1 Kir6.2 channels and pancreatic β-cell KATP channels (EC50 = 7 μM and 0.3-10.3 μM, respectively). VU 0071063 inhibits insulin secretion in pancreatic β-cells by inducing hyperpolarization of β-cell membranes and reduces voltage-dependent calcium channel activation and Ca2+ influx. VU 0071063 is active in vivo and brain penetrant.

Specifications

Chemical Name or Material 7-[[4-(1,1-Dimethylethyl)phenyl]methyl]-3,7-dihydro-1,3-dimethyl-1 H-purine-2,6-dione
CAS 333415-38-6
Quantity 10 mg
Target Inward Rectifier Potassium Channel Activators
Molecular Formula C18H22N4O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.