Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 0463271
GREENER_CHOICESDP

Catalog No. 471950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-195-0 50 mg
47-191-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-195-0 Supplier Tocris Bioscience™ Supplier No. 4719/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective KCC2 inhibitor

VU 0463271 is a potent and selective inhibitor of the neuronal K-Cl cotransporter, KCC2 (IC50 = 61 nM); displays >100-fold selectivity versus the Na-K-2Cl cotransporter 1 (NKCC1) and no activity against a panel of 68 GPCRs, ion channels and transporters.

Specifications

Chemical Name or Material N-Cyclopropyl-N-(4-methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]acetamide
CAS 1391737-01-1
Quantity 50 mg
Target KCC2 Inhibitors
Molecular Formula C19H18N4OS2
Formula Weight 382.5
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.