Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 0542270
SDP

Catalog No. 815150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-515-0 50 mg
81-511-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 81-515-0 Supplier Tocris Bioscience™ Supplier No. 8151/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of Kir6.1 KATP channels

VU 0542270 is a selective inhibitor of Kir6.1/SUR1 KATP channels (IC50 = 104 nM). Shows no activity at other Kir channel types at up to 30 μM. Dose-dependently induces constriction of ductus arteriosus vessels.

Specifications

Chemical Name or Material 1-[(4-Methyl-2-thiophen-2-yl-1,3-thiazol-5-yl)methyl]-3-thiophen-2-ylurea
CAS 1396814-79-1
Quantity 50 mg
Target Inward Rectifier Potassium Channel Activators
Molecular Formula C14H13N3OS3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.